Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053254.2 | NP_444484.1 | 2 - 3 | 185 | 72 | |
GenBank mRNA | AB221587.1 | - | 1 - 2 | 46 | 72 | |
AF145957.2 | - | 2 - 3 | 185 | 72 | ||
AK135768.1 | - | 2 - 3 | 162 | 72 | ||
BC023315.1 | - | 2 - 3 | 119 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |