Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gadd45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008655.1 | NP_032681.1 | 2 - 3 | 376 | 57 | |
GenBank mRNA | AK010420.1 | - | 2 - 3 | 394 | 57 | |
AK149895.1 | - | 2 - 3 | 394 | 57 | ||
AK154952.1 | - | 2 - 3 | 394 | 57 | ||
BC023815.1 | - | 2 - 3 | 388 | 5355150 | 57 | |
X54149.1 | - | 2 - 3 | 376 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |