Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028657.3 | NP_082933.2 | 3 - 4 | 664 | 61 | |
GenBank mRNA | AK149678.1 | - | 3 - 4 | 670 | 61 | |
AK154386.1 | - | 3 - 4 | 666 | 61 | ||
AK154902.1 | - | 3 - 4 | 744 | 61 | ||
AK170185.1 | - | 3 - 4 | 684 | 61 | ||
AK171084.1 | - | 3 - 4 | 666 | 61 | ||
BC118620.1 | - | 3 - 4 | 664 | 40107417 | 61 | |
BC119790.1 | - | 3 - 4 | 664 | 40107416 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |