Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pias4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021501.4 | NP_067476.2 | 5 - 6 | 662 | 63 | |
GenBank mRNA | AF109174.1 | - | 5 - 6 | 864 | 63 | |
AK131822.1 | - | 662 | 63 | |||
AK131903.1 | - | 5 - 6 | 654 | 63 | ||
AK145367.1 | - | 757 | 63 | |||
AK147813.1 | - | 5 - 6 | 693 | 63 | ||
AK148066.1 | - | 664 | 63 | |||
BC020476.1 | - | 4 - 5 | 628 | 63 | ||
BC025159.1 | - | 5 - 6 | 662 | 4236082 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |