Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB093224.1 | - | 9 - 10 | 2776 | 66 | |
AK004685.1 | - | 5 - 6 | 902 | 66 | ||
AK082152.1 | - | 4 - 5 | 665 | 66 | ||
AK147276.1 | - | 14 - 15 | 4136 | 66 | ||
AK147535.1 | - | 15 - 16 | 6254 | 66 | ||
BC040463.1 | - | 5 - 6 | 1203 | 66 | ||
BC058665.1 | - | 11 - 12 | 3021 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |