Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143776.1 | NP_001137248.1 | 9 - 10 | 1180 | 82 | |
NM_001143777.1 | NP_001137249.1 | 10 - 11 | 1284 | 82 | ||
NM_024244.4 | NP_077206.3 | 9 - 10 | 1180 | 82 | ||
GenBank mRNA | AK004797.1 | - | 9 - 10 | 1164 | 82 | |
AK032053.1 | - | 10 - 11 | 1096 | 82 | ||
AK036299.1 | - | 10 - 11 | 1247 | 82 | ||
AK041147.1 | - | 9 - 10 | 1181 | 82 | ||
AK044049.1 | - | 10 - 11 | 1207 | 82 | ||
BC129839.1 | - | 9 - 10 | 1136 | 40060810 | 82 | |
BC129840.1 | - | 9 - 10 | 1136 | 40060811 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |