Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aifm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153779.2 | NP_722474.2 | 8 - 9 | 1071 | 56 | |
GenBank mRNA | BC038129.1 | - | 8 - 9 | 1071 | 1395873 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |