Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145969.1 | NP_001139441.1 | 1 - 2 | 77 | 63 | |
GenBank mRNA | CN836837.1 | - | 1 - 2 | 80 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |