Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_074087.1 | 4 - 5 | 434 | 67 | ||
GenBank mRNA | BE685863.1 | - | 4 - 5 | 226 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |