Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK018269.1 | - | 1 - 2 | 243 | 61 | |
AK046113.1 | - | 1 - 2 | 33 | 61 | ||
BC119094.1 | - | 1 - 2 | 212 | 8733843 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |