Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap19-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130873.1 | NP_570943.1 | 1 | 237 | 88 | |
GenBank mRNA | AF345294.1 | - | 1 | 237 | 88 | |
BC116200.1 | - | 1 | 237 | 40090087 | 88 | |
BC116201.1 | - | 1 | 237 | 40090088 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |