Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033498.1 | NP_001028670.1 | 2 - 3 | 391 | 82 | |
GenBank mRNA | AK040545.1 | - | 3 - 4 | 280 | 82 | |
AK041631.1 | - | 2 - 3 | 391 | 82 | ||
BC138948.1 | - | 2 - 3 | 208 | 8861970 | 82 | |
BC138949.1 | - | 2 - 3 | 208 | 8861971 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |