Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153119.3 | NP_694759.1 | 1 - 2 | 144 | 69 | |
GenBank mRNA | AK028069.1 | - | 1 - 2 | 135 | 69 | |
AK170479.1 | - | 1 - 2 | 128 | 69 | ||
BC028908.1 | - | 1 - 2 | 103 | 3668775 | 69 | |
BY193678.1 | - | 1 - 2 | 144 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |