Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213614.2 | NP_998779.2 | 10 - 11 | 1040 | 84 | |
GenBank mRNA | AF033350.1 | - | 8 - 9 | 887 | 84 | |
AK134700.1 | - | 10 - 11 | 977 | 84 | ||
BC059848.1 | - | 8 - 9 | 918 | 84 | ||
BC079631.1 | - | 4 - 5 | 354 | 84 | ||
BC094347.1 | - | 9 - 10 | 916 | 84 | ||
BC141073.1 | - | 10 - 11 | 1050 | 9055685 | 84 | |
BC145331.1 | - | 10 - 11 | 1050 | 9053887 | 84 | |
BC145332.1 | - | 10 - 11 | 1099 | 9053888 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |