Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290528.1 | NP_001277457.1 | 9 - 10 | 3054 | 95 | |
GenBank mRNA | BC046514.1 | - | 6 - 7 | 1450 | 95 | |
BC057197.1 | - | 8 - 9 | 1763 | 95 | ||
BC078445.1 | - | 9 - 10 | 2093 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Protein Kinase 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons |