Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176954.3 | NP_795928.2 | 10 - 11 | 1170 | 121 | |
GenBank mRNA | AK029796.1 | - | 10 - 11 | 2344 | 121 | |
AK132321.1 | - | 10 - 11 | 1136 | 121 | ||
AK158669.1 | - | 9 - 10 | 947 | 121 | ||
BC079845.1 | - | 10 - 11 | 1141 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |