Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164147.1 | NP_001157619.1 | 15 - 16 | 1720 | 84 | |
NM_001164148.1 | NP_001157620.1 | 15 - 16 | 1717 | 84 | ||
NM_001164149.1 | NP_001157621.1 | 15 - 16 | 1714 | 84 | ||
NM_001164150.1 | NP_001157622.1 | 15 - 16 | 1720 | 84 | ||
NM_001164151.1 | NP_001157623.1 | 15 - 16 | 1717 | 84 | ||
NM_001164152.1 | NP_001157624.1 | 15 - 16 | 1717 | 84 | ||
NM_001164153.1 | NP_001157625.1 | 15 - 16 | 1717 | 84 | ||
NM_011548.4 | NP_035678.3 | 15 - 16 | 1714 | 84 | ||
GenBank mRNA | AF352579.1 | - | 15 - 16 | 1409 | 84 | |
AK017617.1 | - | 15 - 16 | 1682 | 84 | ||
AK034777.1 | - | 15 - 16 | 1700 | 84 | ||
AK038738.1 | - | 15 - 16 | 1704 | 84 | ||
AK146786.1 | - | 15 - 16 | 1712 | 84 | ||
AK147735.1 | - | 14 - 15 | 1377 | 84 | ||
AK156265.1 | - | 15 - 16 | 84 | |||
AK161009.1 | - | 15 - 16 | 1706 | 84 | ||
AK170674.1 | - | 15 - 16 | 1712 | 84 | ||
BC006860.1 | - | 15 - 16 | 1668 | 2631291 | 84 | |
BC018260.1 | - | 14 - 15 | 1342 | 4187386 | 84 | |
D16631.1 | - | 1 - 2 | 169 | 84 | ||
D16632.1 | - | 1 - 2 | 169 | 84 | ||
D16633.1 | - | 1 - 2 | 169 | 84 | ||
D16634.1 | - | 1 - 2 | 169 | 84 | ||
D16635.1 | - | 1 - 2 | 172 | 84 | ||
D16636.1 | - | 1 - 2 | 169 | 84 | ||
D16637.1 | - | 1 - 2 | 169 | 84 | ||
D16638.1 | - | 1 - 2 | 169 | 84 | ||
X17500.1 | - | 2 - 3 | 222 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |