Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164147.1 | NP_001157619.1 | 1 - 2 | 136 | ||
NM_001164148.1 | NP_001157620.1 | 1 - 2 | 136 | |||
NM_001164149.1 | NP_001157621.1 | 1 - 2 | 136 | |||
NM_001164150.1 | NP_001157622.1 | 1 - 2 | 136 | |||
NM_001164151.1 | NP_001157623.1 | 1 - 2 | 136 | |||
NM_001164152.1 | NP_001157624.1 | 1 - 2 | 136 | |||
NM_001164153.1 | NP_001157625.1 | 1 - 2 | 136 | |||
NM_011548.4 | NP_035678.3 | 1 - 2 | 136 | |||
GenBank mRNA | AK017617.1 | - | 1 - 2 | 135 | ||
AK034777.1 | - | 1 - 2 | 136 | |||
AK038738.1 | - | 1 - 2 | 136 | |||
AK146786.1 | - | 1 - 2 | 136 | |||
AK156265.1 | - | 1 - 2 | 320 | 136 | ||
AK161009.1 | - | 1 - 2 | 136 | |||
AK170674.1 | - | 1 - 2 | 136 | |||
BC006860.1 | - | 1 - 2 | 291 | 2631291 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |