Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028049.2 | NP_082325.2 | 2 - 3 | 394 | 68 | |
GenBank mRNA | AK002745.1 | - | 2 - 3 | 274 | 68 | |
AK005473.1 | - | 1 - 2 | 222 | 68 | ||
BC018273.1 | - | 2 - 3 | 334 | 4458296 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |