Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600006645.mm.1 |
NM_001252450.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006645.mm.1 |
NM_001252450.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006645.mm.1 |
NM_001252451.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006645.mm.1 |
NM_001252451.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006645.mm.1 |
NM_171826.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006645.mm.1 |
NM_171826.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006645.mm.1 |
NR_045517.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006645.mm.1 |
NR_045517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006645.mm.1 |
NR_045518.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006645.mm.1 |
NR_045518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006645.mm.1 |
TC1600000765.mm.1 |
MTA TC |