Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008319.2 | NP_032345.2 | 3 - 4 | 769 | 113 | |
GenBank mRNA | AK135040.1 | - | 3 - 4 | 682 | 113 | |
BC113140.1 | - | 3 - 4 | 717 | 40048010 | 113 | |
U06483.1 | - | 3 - 4 | 717 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |