Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042707.2 | NP_001036172.1 | 16 - 17 | 2334 | 60 | |
NM_001042708.2 | NP_001036173.1 | 17 - 18 | 2373 | 60 | ||
NM_001042709.2 | NP_001036174.1 | 16 - 17 | 2334 | 60 | ||
NM_001277321.1 | NP_001264250.1 | 17 - 18 | 2373 | 60 | ||
NM_001277322.1 | NP_001264251.1 | 18 - 19 | 2301 | 60 | ||
NM_010561.3 | NP_034691.2 | 17 - 18 | 2373 | 60 | ||
GenBank mRNA | AB221609.1 | - | 15 - 16 | 2049 | 60 | |
AF098967.1 | - | 18 - 19 | 2250 | 60 | ||
AF497751.2 | - | 17 - 18 | 2241 | 60 | ||
AF497752.2 | - | 16 - 17 | 2202 | 60 | ||
AK048096.1 | - | 16 - 17 | 2264 | 60 | ||
AK088604.1 | - | 3 - 4 | 367 | 60 | ||
AK128936.1 | - | 17 - 18 | 2306 | 60 | ||
AK153875.1 | - | 3 - 4 | 368 | 60 | ||
BC047272.1 | - | 17 - 18 | 2293 | 6440312 | 60 | |
BC060599.1 | - | 14 - 15 | 1939 | 60 | ||
DQ104405.1 | - | 17 - 18 | 2241 | 60 | ||
DQ104406.1 | - | 16 - 17 | 2202 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |