Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153542.1 | NP_705770.1 | 3 - 4 | 360 | 87 | |
GenBank mRNA | AK136380.1 | - | 3 - 4 | 387 | 87 | |
AK139384.1 | - | 3 - 4 | 393 | 87 | ||
AK145920.1 | - | 3 - 4 | 377 | 87 | ||
AK150142.1 | - | 3 - 4 | 361 | 87 | ||
AK150246.1 | - | 3 - 4 | 361 | 87 | ||
BC036304.1 | - | 3 - 4 | 360 | 3968534 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |