Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010421.4 | NP_034551.2 | 2 - 3 | 404 | 91 | |
GenBank mRNA | AK075895.1 | - | 2 - 3 | 380 | 91 | |
AK075911.1 | - | 2 - 3 | 405 | 91 | ||
AK080777.1 | - | 2 - 3 | 377 | 91 | ||
AK144168.1 | - | 2 - 3 | 532 | 91 | ||
AK150444.1 | - | 2 - 3 | 375 | 91 | ||
AK151960.1 | - | 2 - 3 | 379 | 91 | ||
AK159091.1 | - | 2 - 3 | 396 | 91 | ||
AK159814.1 | - | 2 - 3 | 390 | 91 | ||
AK159890.1 | - | 2 - 3 | 377 | 91 | ||
AK160112.1 | - | 2 - 3 | 364 | 91 | ||
AK167013.1 | - | 2 - 3 | 390 | 91 | ||
AK170903.1 | - | 2 - 3 | 390 | 91 | ||
BC010755.1 | - | 2 - 3 | 373 | 4225233 | 91 | |
U07631.1 | - | 2 - 3 | 396 | 91 | ||
X64331.1 | - | 2 - 3 | 347 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |