Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024139.1 | NP_001019310.1 | 2 - 3 | 1301 | 88 | |
GenBank mRNA | BC094677.1 | - | 2 - 3 | 1301 | 30619053 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |