Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011176.4 | NP_035306.2 | 2 - 3 | 412 | 79 | |
GenBank mRNA | AF042822.3 | - | 2 - 3 | 301 | 79 | |
AK052738.1 | - | 2 - 3 | 412 | 79 | ||
AK085410.1 | - | 2 - 3 | 410 | 79 | ||
AK166745.1 | - | 2 - 3 | 403 | 79 | ||
BC005496.1 | - | 2 - 3 | 411 | 3488059 | 79 | |
BC008514.1 | - | 2 - 3 | 411 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |