Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080817.1 | NP_001074286.1 | 7 - 8 | 989 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |