Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfrkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172766.3 | NP_766354.2 | 3 - 4 | 460 | 72 | |
GenBank mRNA | AK041206.1 | - | 3 - 4 | 456 | 72 | |
BC033595.1 | - | 3 - 4 | 450 | 4947357 | 72 | |
BC059861.1 | - | 3 - 4 | 439 | 72 | ||
BC060076.1 | - | 3 - 4 | 447 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |