Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700024785.mm.1 |
TC0700002776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024785.mm.1 |
NM_001168509.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024785.mm.1 |
NM_001168509.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024785.mm.1 |
NM_001168510.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024785.mm.1 |
NM_001168510.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024785.mm.1 |
NM_001168511.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024785.mm.1 |
NM_001168511.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024785.mm.1 |
NM_001168512.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024785.mm.1 |
NM_001168512.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024785.mm.1 |
NM_146187.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024785.mm.1 |
NM_146187.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024790.mm.1 |
TC0700002776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024790.mm.1 |
NM_001168509.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024790.mm.1 |
NM_001168509.1 |
Refseq |