Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026815.2 | NP_081091.1 | 3 - 4 | 311 | 64 | |
GenBank mRNA | AF262335.1 | - | 2 - 3 | 283 | 64 | |
AK004014.1 | - | 3 - 4 | 312 | 64 | ||
BC132047.1 | - | 2 - 3 | 303 | 40130324 | 64 | |
BC138390.1 | - | 2 - 3 | 303 | 8861410 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |