Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019459.2 | NP_062332.2 | 13 - 14 | 1885 | 93 | |
GenBank mRNA | AB513652.1 | - | 13 - 14 | 1757 | 93 | |
AF168466.1 | - | 13 - 14 | 1757 | 93 | ||
AF172256.1 | - | 13 - 14 | 1885 | 93 | ||
AF191090.1 | - | 13 - 14 | 2213 | 93 | ||
AK141081.1 | - | 13 - 14 | 1908 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |