Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25031675 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25031676 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1996479 |
NM_019459.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
27074 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
184456 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
796270 |
AK008079.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
796270 |
AK018567.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
796270 |
BC038900.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
796270 |
BC057172.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
796270 |
NM_023256.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
158507 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
806934 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
481315 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
217821 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
419089 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
284675 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
860500 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
19011 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
365451 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
520018 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
530603 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
775591 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
371344 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
555590 |
AK016427.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
290190 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
108479 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
154096 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
100616 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
47403 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P128170 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P170699 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1996479 |
NM_019459.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P496741 |
AF172256.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P496741 |
AF191090.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P496741 |
AK141081.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P496741 |
AF168466.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P496741 |
AB513652.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031651 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031654 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031636 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031646 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031641 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031658 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031655 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031653 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031638 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031649 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031644 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031647 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031637 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031640 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25031639 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01493116 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P128170 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06154634 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06154716 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06154612 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P170699 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06154674 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P496741 |
AF172256.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P496741 |
AF191090.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P496741 |
AK141081.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P496741 |
AF168466.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P496741 |
AB513652.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P118763 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P128170 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P170699 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P06154634 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05890679 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05890689 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05890695 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25031676 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25031675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031595 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031808 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031602 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031678 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031634 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031738 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031799 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031663 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031786 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031784 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25031694 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06102491 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031626 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031640 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06549257 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031777 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031682 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031669 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031676 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031766 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031672 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031684 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031663 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031674 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031755 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031661 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031659 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031683 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031653 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031667 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031776 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031687 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031688 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06471120 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031662 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031770 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031678 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031651 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031641 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05890678 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031666 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06549256 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06409537 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06471119 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031629 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031647 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031673 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031654 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06266066 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031764 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06620198 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031762 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031630 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031625 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031644 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031774 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05890690 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06506078 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06506083 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031649 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031677 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031751 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031760 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031658 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031634 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06549258 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031636 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031750 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031778 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06009903 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25031638 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06560827 |
- |
- |