Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kirrel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172898.3 | NP_766486.1 | 2 - 3 | 868 | 77 | |
GenBank mRNA | AK049284.1 | - | 2 - 3 | 869 | 77 | |
AY305303.1 | - | 1 - 2 | 63 | 77 | ||
BC052773.1 | - | 2 - 3 | 331 | 6314924 | 77 | |
BC158021.1 | - | 2 - 3 | 376 | 9007327 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |