Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028047.2 | NP_082323.1 | 8 - 9 | 1180 | 67 | |
GenBank mRNA | AK005126.1 | - | 8 - 9 | 1180 | 67 | |
AK039368.1 | - | 8 - 9 | 1179 | 67 | ||
AK141576.1 | - | 8 - 9 | 1171 | 67 | ||
AK169651.1 | - | 8 - 9 | 1104 | 67 | ||
BC034211.1 | - | 8 - 9 | 1095 | 4988361 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |