Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007691.5 | NP_031717.2 | 2 - 3 | 179 | 96 | |
GenBank mRNA | AF016583.1 | - | 1 - 2 | 88 | 96 | |
AK011258.1 | - | 2 - 3 | 123 | 96 | ||
AK033179.1 | - | 2 - 3 | 149 | 96 | ||
AK045336.1 | - | 2 - 3 | 121 | 96 | ||
AK166675.1 | - | 2 - 3 | 180 | 96 | ||
BC053394.1 | - | 2 - 3 | 115 | 96 | ||
BC064445.1 | - | 1 - 2 | 579 | 96 | ||
BC100386.1 | - | 1 - 2 | 254 | 30481390 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |