Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008408.4 | NP_032434.3 | 5 - 6 | 488 | 72 | |
GenBank mRNA | AK030363.1 | - | 5 - 6 | 552 | 72 | |
AK077877.1 | - | 5 - 6 | 512 | 72 | ||
AK081547.1 | - | 5 - 6 | 544 | 72 | ||
AK129027.1 | - | 5 - 6 | 526 | 72 | ||
AK150640.1 | - | 5 - 6 | 533 | 72 | ||
AK151541.1 | - | 5 - 6 | 544 | 72 | ||
AK151791.1 | - | 5 - 6 | 544 | 72 | ||
AK152242.1 | - | 5 - 6 | 533 | 72 | ||
AK153840.1 | - | 5 - 6 | 489 | 72 | ||
AK153886.1 | - | 5 - 6 | 589 | 72 | ||
AK160037.1 | - | 5 - 6 | 527 | 72 | ||
AK160607.1 | - | 5 - 6 | 489 | 72 | ||
AK168505.1 | - | 5 - 6 | 533 | 72 | ||
AK168937.1 | - | 5 - 6 | 533 | 72 | ||
AK169669.1 | - | 5 - 6 | 512 | 72 | ||
AX799090.1 | - | 5 - 6 | 531 | 72 | ||
BC028897.1 | - | 4 - 5 | 439 | 72 | ||
BC037612.1 | - | 5 - 6 | 527 | 5353797 | 72 | |
BC085313.1 | - | 5 - 6 | 517 | 30443880 | 72 | |
L34260.1 | - | 5 - 6 | 531 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |