Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199222.3 | NP_954692.2 | 1 - 2 | 279 | 64 | |
GenBank mRNA | AB211986.1 | - | 1 - 2 | 200 | 64 | |
BC020188.1 | - | 1 - 2 | 290 | 4925160 | 64 | |
BY708954.1 | - | 1 - 2 | 280 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |