Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148941.2 | NP_683743.2 | 4 - 5 | 806 | 102 | |
GenBank mRNA | AF277093.1 | - | 4 - 5 | 602 | 102 | |
AK029065.1 | - | 4 - 5 | 667 | 102 | ||
AK030666.1 | - | 4 - 5 | 755 | 102 | ||
BC037030.1 | - | 4 - 5 | 749 | 4503493 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |