Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153112.3 | NP_694752.1 | 2 - 3 | 283 | 68 | |
GenBank mRNA | AK143204.1 | - | 2 - 3 | 284 | 68 | |
AY059394.1 | - | 2 - 3 | 207 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |