Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172134.2 | NP_742146.1 | 2 - 3 | 240 | 68 | |
GenBank mRNA | AK039194.1 | - | 2 - 3 | 222 | 68 | |
AK080846.1 | - | 2 - 3 | 216 | 68 | ||
AK145470.1 | - | 2 - 3 | 204 | 68 | ||
AK166078.1 | - | 2 - 3 | 223 | 68 | ||
AK166464.1 | - | 2 - 3 | 205 | 68 | ||
BC027745.1 | - | 2 - 3 | 277 | 5064695 | 68 | |
BC032171.1 | - | 2 - 3 | 211 | 68 | ||
BI143426.1 | - | 2 - 3 | 269 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |