Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144784.3 | NP_659033.1 | 3 - 4 | 302 | 84 | |
GenBank mRNA | AJ510150.1 | - | 3 - 4 | 294 | 84 | |
AK032097.1 | - | 3 - 4 | 273 | 84 | ||
AK075593.1 | - | 3 - 4 | 272 | 84 | ||
AK081715.1 | - | 3 - 4 | 302 | 84 | ||
AK146241.1 | - | 3 - 4 | 257 | 84 | ||
AK163402.1 | - | 3 - 4 | 306 | 84 | ||
AK169771.1 | - | 3 - 4 | 295 | 84 | ||
BC024429.1 | - | 3 - 4 | 305 | 84 | ||
BC024763.1 | - | 3 - 4 | 274 | 5365469 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |