Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081688.2 | NP_001075157.2 | 14 - 15 | 2567 | 81 | |
GenBank mRNA | BC158102.1 | - | 14 - 15 | 2663 | 9007387 | 81 |
BC172169.1 | - | 14 - 15 | 2663 | 9054839 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |