Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700003575.mm.1 |
TC0700004617.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003575.mm.1 |
NM_133770.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003575.mm.1 |
NM_133770.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003577.mm.1 |
TC0700004617.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003577.mm.1 |
NM_133770.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003577.mm.1 |
NM_133770.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003574.mm.1 |
TC0700004617.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003574.mm.1 |
NM_133770.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003574.mm.1 |
NM_133770.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003569.mm.1 |
TC0700004617.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003569.mm.1 |
NM_133770.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003569.mm.1 |
NM_133770.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003573.mm.1 |
TC0700004617.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003573.mm.1 |
NM_133770.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003573.mm.1 |
NM_133770.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003572.mm.1 |
TC0700004617.mm.1 |
MTA TC |