Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133770.2 | NP_598531.2 | 7 - 8 | 738 | 68 | |
GenBank mRNA | AK002619.1 | - | 7 - 8 | 729 | 68 | |
BC013485.1 | - | 4 - 5 | 301 | 4242474 | 68 | |
BC083324.1 | - | 6 - 7 | 681 | 6404832 | 68 | |
BC093498.1 | - | 6 - 7 | 675 | 3669346 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |