Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019727.2 | NP_062701.2 | 9 - 10 | 1039 | 95 | |
GenBank mRNA | AB019214.1 | - | 9 - 10 | 979 | 95 | |
AF154120.1 | - | 9 - 10 | 1043 | 95 | ||
AK149850.1 | - | 9 - 10 | 951 | 95 | ||
AK154079.1 | - | 9 - 10 | 952 | 95 | ||
AK155746.1 | - | 9 - 10 | 927 | 95 | ||
AK161241.1 | - | 9 - 10 | 1020 | 95 | ||
AK167177.1 | - | 9 - 10 | 994 | 95 | ||
AK167993.1 | - | 9 - 10 | 934 | 95 | ||
AK168742.1 | - | 9 - 10 | 1032 | 95 | ||
AK192993.1 | - | 3 - 4 | 229 | 95 | ||
BC066044.1 | - | 9 - 10 | 996 | 6807363 | 95 | |
BC066189.1 | - | 9 - 10 | 930 | 30471747 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |