Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009022.4 | NP_033048.2 | 2 - 3 | 277 | 63 | |
GenBank mRNA | AK078553.1 | - | 2 - 3 | 218 | 63 | |
BC075704.1 | - | 2 - 3 | 244 | 30471325 | 63 | |
CJ166631.1 | - | 2 - 3 | 278 | 63 | ||
X99273.1 | - | 2 - 3 | 222 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |