Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178602.3 | NP_848717.1 | 1 - 2 | 295 | 58 | |
GenBank mRNA | AK014316.1 | - | 1 - 2 | 227 | 58 | |
AK017773.1 | - | 1 - 2 | 238 | 58 | ||
AK051868.1 | - | 1 - 2 | 229 | 58 | ||
AK052037.1 | - | 1 - 2 | 249 | 58 | ||
AK078750.1 | - | 1 - 2 | 229 | 58 | ||
AK079119.1 | - | 1 - 2 | 213 | 58 | ||
AK136806.1 | - | 1 - 2 | 244 | 58 | ||
AK151221.1 | - | 1 - 2 | 232 | 58 | ||
AK159924.1 | - | 1 - 2 | 242 | 58 | ||
AK166738.1 | - | 1 - 2 | 242 | 58 | ||
BC018502.1 | - | 1 - 2 | 221 | 2649742 | 58 | |
BC062199.1 | - | 1 - 2 | 215 | 4013916 | 58 | |
BQ443340.1 | - | 1 - 2 | 311 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |