Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019458.3 | NP_062331.2 | 8 - 9 | 849 | 76 | |
GenBank mRNA | AK017762.1 | - | 8 - 9 | 919 | 76 | |
AK134857.1 | - | 8 - 9 | 862 | 76 | ||
AK148538.1 | - | 8 - 9 | 884 | 76 | ||
AK151665.1 | - | 8 - 9 | 895 | 76 | ||
AK152778.1 | - | 8 - 9 | 865 | 76 | ||
AK152836.1 | - | 8 - 9 | 895 | 76 | ||
BC010317.1 | - | 7 - 8 | 893 | 76 | ||
BC029843.1 | - | 8 - 9 | 849 | 2651081 | 76 | |
BC083337.1 | - | 8 - 9 | 830 | 5701517 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |