Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175319.4 | NP_780528.1 | 10 - 11 | 1344 | 79 | |
GenBank mRNA | AK049131.1 | - | 10 - 11 | 1345 | 79 | |
BC094908.1 | - | 10 - 11 | 1240 | 30021730 | 79 | |
BC132375.1 | - | 10 - 11 | 1331 | 40130652 | 79 | |
BC138307.1 | - | 10 - 11 | 1330 | 8861327 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |