Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023831.3 | NP_076320.2 | 7 - 8 | 769 | 70 | |
GenBank mRNA | AJ249981.1 | - | 7 - 8 | 813 | 70 | |
AK005360.1 | - | 7 - 8 | 777 | 70 | ||
AK032880.1 | - | 7 - 8 | 770 | 70 | ||
BC010326.1 | - | 6 - 7 | 568 | 3599905 | 70 | |
BC080764.1 | - | 7 - 8 | 711 | 6488976 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |